Stampa…

1-Bromo-2-(methylsulphinyl)benzene ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1-Bromo-2-(methylsulphinyl)benzene, 2-Bromophenyl methyll sulphoxide

B4990-1G B4990-5G
TCIAB4990-1GEA 81 EUR
TCIAB4990-1G TCIAB4990-5G
1-Bromo-2-(methylsulphinyl)benzene ≥98.0% (by GC)
1-Bromo-2-(methylsulphinyl)benzene
Formula: C₇H₇BrOS
Peso molecolare: 219,11 g/mol
Punto di ebollizione: 109°C
MDL Number: MFCD18089235
CAS Number: 7321-58-6

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR