Stampa…

Bis(2-bromophenyl)acetylene ≥96.0% (by GC)

Fornitore: TCI EUROPE

B4989-1G B4989-5G
TCIAB4989-1GEA 152 EUR
TCIAB4989-1G TCIAB4989-5G
Bis(2-bromophenyl)acetylene ≥96.0% (by GC)
Bis(2-bromophenyl)acetylene
Formula: C₁₄H₈Br₂
Peso molecolare: 336,03 g/mol
MDL Number: MFCD00238712
CAS Number: 38399-13-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Melting point 79.0 to 83.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR