Stampa…

2-Cyano-5-fluorobenzyl bromide ≥98.0% (by GC)

Fornitore: TCI
Pericolo

B4988-1G B4988-5G
TCIAB4988-1GEA 33 EUR
TCIAB4988-1G TCIAB4988-5G
2-Cyano-5-fluorobenzyl bromide ≥98.0% (by GC)
2-Cyano-5-fluorobenzyl bromide
Formula: C₈H₅BrFN
MDL Number: MFCD08059542
CAS Number: 421552-12-7
UN: 3261
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Melting point 67.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR