Stampa…

3-Bromo-5-nitrobenzaldehyde ≥96.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B4982-1G B4982-5G
TCIAB4982-1GEA 36 EUR
TCIAB4982-1G TCIAB4982-5G
3-Bromo-5-nitrobenzaldehyde ≥96.0% (by GC)
3-Bromo-5-nitrobenzaldehyde
Formula: C₇H₄BrNO₃
Peso molecolare: 230,02 g/mol
MDL Number: MFCD00100097
CAS Number: 355134-13-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 100.0 to 104.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR