9,9-Bis[4-[N-(1-naphthyl)anilino]phenyl]fluorene ≥97.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
B4979-200MG
B4979-1G
TCIAB4979-200MGEA
83
EUR
TCIAB4979-200MG
TCIAB4979-1G
9,9-Bis[4-[N-(1-naphthyl)anilino]phenyl]fluorene ≥97.0% (by HPLC, total nitrogen)
9,9-Bis[4-[N-(1-naphthyl)anilino]phenyl]fluorene
Formula:
C₅₇H₄₀N₂ Peso molecolare: 752,96 g/mol |
CAS Number:
510775-24-3 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...