Stampa…

4,4'-Bis(4,6-diphenyl-1,3,5-triazin-2-yl)biphenyl ≥95.0% (by HPLC, total nitrogen)

Fornitore: TCI

B4977-200MG B4977-1G
TCIAB4977-200MGEA 80 EUR
TCIAB4977-200MG TCIAB4977-1G
4,4'-Bis(4,6-diphenyl-1,3,5-triazin-2-yl)biphenyl ≥95.0% (by HPLC, total nitrogen)
4,4'-Bis(4,6-diphenyl-1,3,5-triazin-2-yl)biphenyl

This product is unavailable in the U.S.

Formula: C₄₂H₂₈N₆
MDL Number: MFCD28291963
CAS Number: 266349-83-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light gray to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR