Stampa…

4-Bromo-2,6-pyridinedicarboxylic acid monohydrate ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Bromopyridine-2,6-dicarboxylic acid

B4970-200MG B4970-1G
TCIAB4970-200MGEA 100 EUR
TCIAB4970-200MG TCIAB4970-1G
4-Bromo-2,6-pyridinedicarboxylic acid monohydrate ≥98.0% (by GC, titration analysis)
4-Bromo-2,6-pyridinedicarboxylic acid monohydrate
Formula: C₇H₄BrNO₄·H₂O
Peso molecolare: 246,02 g/mol
MDL Number: MFCD09033573
CAS Number: 162102-81-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Water 5.0 to 9.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR