Stampa…

9,10-Bis[N,N-di(p-tolyl)amino]anthracene ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: N9,N9,N10,N10-Tetra-p-tolylanthracene-9,10-diamine, N,N,N',N'-Tetrakis(4-methylphenyl)-9,10-anthracenediamine

B4966-200MG B4966-1G
TCIAB4966-200MGEA 100 EUR
TCIAB4966-200MG TCIAB4966-1G
9,10-Bis[N,N-di(p-tolyl)amino]anthracene ≥98.0% (by HPLC)
9,10-Bis[N,N-di(p-tolyl)amino]anthracene
Formula: C₄₂H₃₆N₂
Peso molecolare: 568,76 g/mol
MDL Number: MFCD12022465
CAS Number: 177799-16-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5faff8871636a58b4c542fb988267edd4dc6fa1f&vid=5faff8871636a58b4c542fb988267edd4dc6fa1f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR