Stampa…

9-Bromo-7,7-dimethyl-7H-benzo[c]fluorene ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B4952-200MG B4952-1G
TCIAB4952-200MGEA 57 EUR
TCIAB4952-200MG TCIAB4952-1G
9-Bromo-7,7-dimethyl-7H-benzo[c]fluorene ≥98.0% (by GC)
9-Bromo-7,7-dimethyl-7H-benzo[c]fluorene
Formula: C₁₉H₁₅Br
Peso molecolare: 323,23 g/mol
MDL Number: MFCD27980277
CAS Number: 1198396-46-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR