Stampa…

4-(trans-4-Butylcyclohexyl)cyclohexanone ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: trans-[4'-Butyl-1,1'-bicyclohexyl]-4-one

B4946-5G B4946-25G
TCIAB4946-5GEA 37 EUR
TCIAB4946-5G TCIAB4946-25G
4-(trans-4-Butylcyclohexyl)cyclohexanone ≥98.0% (by GC)
4-(trans-4-Butylcyclohexyl)cyclohexanone
Formula: C₁₆H₂₈O
Peso molecolare: 236,40 g/mol
MDL Number: MFCD06797514
CAS Number: 92413-47-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR