Stampa…

4-Bromo-2,6-diphenylpyrimidine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B4943-200MG B4943-1G
TCIAB4943-200MGEA 30 EUR
TCIAB4943-200MG TCIAB4943-1G
4-Bromo-2,6-diphenylpyrimidine ≥98.0% (by GC)
4-Bromo-2,6-diphenylpyrimidine
Formula: C₁₆H₁₁BrN₂
Peso molecolare: 311,18 g/mol
MDL Number: MFCD00234885
CAS Number: 40734-24-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 110.0 to 114.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR