Stampa…

2,4-Bis(4-biphenylyl)-6-chloro-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,4-Di([1,1'-biphenyl]-4-yl)-6-chloro-1,3,5-triazine

B4941-200MG B4941-1G
TCIAB4941-200MGEA 52 EUR
TCIAB4941-200MG TCIAB4941-1G
2,4-Bis(4-biphenylyl)-6-chloro-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2,4-Bis(4-biphenylyl)-6-chloro-1,3,5-triazine
Formula: C₂₇H₁₈ClN₃
Peso molecolare: 419,91 g/mol
MDL Number: MFCD27979765
CAS Number: 182918-13-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 215.0 to 219.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR