Fmoc-Lys(Fmoc)-OH ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
Synonyms:
Nalpha,Nepsilon-Bis[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine, Fmoc-Lys(Fmoc)-OH, Nalpha,Nepsilon-Di-Fmoc-L-lysine
Formula:
C₃₆H₃₄N₂O₆ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00080275 CAS Number: 78081-87-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 133.0 to 137.0 deg-C |
Specific rotation [a]20/D | -6.0 to -8.0 deg (C=1,DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...