1-(4-Bromophenyl)-2-phenylbenzimidazole ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
1-(4-Bromophenyl)-2-phenyl-1H-benzo[d]imidazole
B4929-1G
B4929-5G
TCIAB4929-1GEA
62
EUR
TCIAB4929-1G
TCIAB4929-5G
1-(4-Bromophenyl)-2-phenylbenzimidazole ≥98.0% (by GC, titration analysis)
1-(4-Bromophenyl)-2-phenylbenzimidazole
Formula:
C₁₉H₁₃BrN₂ Peso molecolare: 349,23 g/mol |
MDL Number:
MFCD09746342 CAS Number: 760212-58-6 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 135.0 to 139.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...