Stampa…

trans-4-(4-Butylcyclohexyl)benzonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 4-(trans-4-Butylcyclohexyl)benzonitrile, 4-(trans-4-Butylcyclohexyl)-1-cyanobenzene

B4924-5G B4924-25G
TCIAB4924-5GEA 26 EUR
TCIAB4924-5G TCIAB4924-25G
trans-4-(4-Butylcyclohexyl)benzonitrile ≥98.0% (by GC)
trans-4-(4-Butylcyclohexyl)benzonitrile
Formula: C₁₇H₂₃N
Peso molecolare: 241,3700 g/mol
MDL Number: MFCD00799425
CAS Number: 61204-00-0
EINECS: 262-655-0
UN: 3439
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR