Stampa…

4-Benzyl-2-hydroxymorpholin-3-one ≥98.0% (by GC)

Fornitore: TCI
Attenzione

B4918-1G B4918-5G
TCIAB4918-1GEA 20 EUR
TCIAB4918-1G TCIAB4918-5G
4-Benzyl-2-hydroxymorpholin-3-one ≥98.0% (by GC)
4-Benzyl-2-hydroxymorpholin-3-one
Formula: C₁₁H₁₃NO₃
MDL Number: MFCD07776435
CAS Number: 287930-73-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 134.0 to 138.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR