Stampa…

7-Bromo-2H-1,4-benzothiazin-3(4H)-one ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 7-Bromo-2H-benzo[b][1,4]thiazin-3(4H)-one

B4902-1G B4902-5G
TCIAB4902-1GEA 71 EUR
TCIAB4902-1G TCIAB4902-5G
7-Bromo-2H-1,4-benzothiazin-3(4H)-one ≥98.0% (by GC)
7-Bromo-2H-1,4-benzothiazin-3(4H)-one
Formula: C₈H₆BrNOS
Peso molecolare: 244,11 g/mol
MDL Number: MFCD02660583
CAS Number: 90814-91-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 212.0 to 216.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR