Stampa…

5-Bromo-2-(5-bromo-2-thienyl)pyridine ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: 5-Bromo-2-(5-bromothiophen-2-yl)pyridine, 2-Bromo-5-(5-bromo-2-pyridyl)thiophene

B4886-200MG B4886-1G
TCIAB4886-200MGEA 101 EUR
TCIAB4886-200MG TCIAB4886-1G
5-Bromo-2-(5-bromo-2-thienyl)pyridine ≥98.0% (by GC)
5-Bromo-2-(5-bromo-2-thienyl)pyridine
Formula: C₉H₅Br₂NS
MDL Number: MFCD29765747
CAS Number: 136902-53-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR