Methyl undecanoate, Supelco®
Fornitore: Merck
Synonyms:
Undecanoic acid methyl ester, Undecylic acid methyl ester
M-Clarity™ quality level = MQ100
94118-1ML
94118-5ML
94118-1MLEA
52.5
EUR
94118-1ML
94118-5ML
Methyl undecanoate, Supelco®
Methyl undecanoate
Formula:
C₁₂H₂₄O₂ Peso molecolare: 200,32 g/mol |
MDL Number:
MFCD00008957 CAS Number: 1731-86-8 EINECS: 217-053-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...