Stampa…

5-Bromo-2-(2-thienyl)pyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Bromo-2-(thiophen-2-yl)pyridine

B4885-200MG B4885-1G
TCIAB4885-200MGEA 60 EUR
TCIAB4885-200MG TCIAB4885-1G
5-Bromo-2-(2-thienyl)pyridine ≥98.0% (by GC, titration analysis)
5-Bromo-2-(2-thienyl)pyridine
Formula: C₉H₆BrNS
Peso molecolare: 240,12 g/mol
MDL Number: MFCD09037419
CAS Number: 91891-74-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 79.0 to 83.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR