Benzyl-4-hydroxypiperidine-1-carboxylate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-Benzyloxycarbonyl-4-hydroxypiperidine, 4-Hydroxy-1-piperidinecarboxylic acid Benzyl ester, 1-Cbz-4-hydroxypiperidine, 1-Carbobenzoxy-4-piperidinol, 1-Carbobenzoxy-4-hydroxypiperidine, Benzyl 4-Hydroxy-1-piperidinecarboxylate, 1-Benzyloxycarbonyl-4-piperidinol, 1-Cbz-4-piperidinol
Formula:
C₁₃H₁₇NO₃ Peso molecolare: 235,29 g/mol Punto di ebollizione: 167°C |
MDL Number:
MFCD01863722 CAS Number: 95798-23-5 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...