Stampa…

N-Boc-S-trityl-L-cysteine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: N-Boc-S-trityl-L-cysteine, N-(tert-Butoxycarbonyl)-S-trityl-L-cysteine, BOC-S-trityl-L-cysteine, (R)-2-((tert-Butoxycarbonyl)amino)-3-(tritylthio)propanoic acid, Boc-Cys(Trt)-OH

B4862-5G B4862-25G
TCIAB4862-5GEA 39 EUR
TCIAB4862-5G TCIAB4862-25G
N-Boc-S-trityl-L-cysteine ≥98.0% (by HPLC, titration analysis)
N-Boc-S-trityl-L-cysteine
Formula: C₂₇H₂₉NO₄S
Peso molecolare: 463,592 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00038251
CAS Number: 21947-98-8
EINECS: 244-674-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +28.0 to +31.0 deg (C=1,EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR