N-Boc-D-serine methyl ester ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
Synonyms:
N-(tert-Butoxycarbonyl)-D-serine methyll ester, Boc-D-Ser-OMe, N-Boc-D-serine methyll ester
B4859-1G
B4859-5G
TCIAB4859-1GEA
16
EUR
TCIAB4859-1G
TCIAB4859-5G
N-Boc-D-serine methyl ester ≥98.0% (by HPLC, total nitrogen)
N-Boc-D-serine methyl ester
Formula:
C₉H₁₇NO₅ Peso molecolare: 219,237 g/mol |
MDL Number:
MFCD00270516 CAS Number: 95715-85-8 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Specific rotation [a]20/D | +15.0 to +21.0 deg (C=5, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...