BAY 11-7085 ≥97.0% (by total nitrogen basis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(E)-3-(4-tert-Butylphenylsulphonyl)acrylonitrile, (E)-3-((4-(tert-Butyl)phenyl)sulphonyl)acrylonitrile
B4845-10MG
B4845-100MG
TCIAB4845-10MGEA
73
EUR
TCIAB4845-10MG
TCIAB4845-100MG
BAY 11-7085 ≥97.0% (by total nitrogen basis)
BAY 11-7085
Formula:
C₁₃H₁₅NO₂S Peso molecolare: 249,328 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD01862602 CAS Number: 196309-76-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 82.0 to 86.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...