Stampa…

4-Bromo-2-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

B4843-1G B4843-5G
TCIAB4843-1GEA 40 EUR
TCIAB4843-1G TCIAB4843-5G
4-Bromo-2-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)
4-Bromo-2-(trifluoromethyl)benzoic acid
Formula: C₈H₄BrF₃O₂
Peso molecolare: 269,02 g/mol
MDL Number: MFCD07787468
CAS Number: 320-31-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 121.0 to 125.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR