Stampa…

5-Bromo-1H-indole-3-carbaldehyde ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Bromoindole-3-carboxaldehyde, 5-Bromo-3-formylindole

B4840-1G B4840-5G
TCIAB4840-1GEA 20 EUR
TCIAB4840-1G TCIAB4840-5G
5-Bromo-1H-indole-3-carbaldehyde ≥98.0% (by GC)
5-Bromo-1H-indole-3-carbaldehyde
Formula: C₉H₆BrNO
Peso molecolare: 224,06 g/mol
MDL Number: MFCD00152016
CAS Number: 877-03-2
EINECS: 212-884-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 204.0 to 208.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR