3-Bromo-2,5-dichloropyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Bromo-2,5-dichloro-pyridine
B4832-1G
B4832-5G
TCIAB4832-1GEA
67
EUR
TCIAB4832-1G
TCIAB4832-5G
3-Bromo-2,5-dichloropyridine ≥98.0% (by GC)
3-Bromo-2,5-dichloropyridine
Formula:
C₅H₂BrCl₂N Peso molecolare: 226,89 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD06659484 CAS Number: 138006-41-4 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 37.0 to 41.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...