4-Bromo-4'-phenyltriphenylamine ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
N-(4-Bromophenyl)-N-phenyl-[1,1'-biphenyl]-4-amine
B4812-1G
B4812-5G
TCIAB4812-1GEA
83
EUR
TCIAB4812-1G
TCIAB4812-5G
4-Bromo-4'-phenyltriphenylamine ≥98.0% (by GC)
4-Bromo-4'-phenyltriphenylamine
Formula:
C₂₄H₁₈BrN Peso molecolare: 400,32 g/mol |
MDL Number:
MFCD24387484 CAS Number: 503299-24-9 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...