Stampa…

3-Bromo-6,9-diphenylcarbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Bromo-6,9-diphenyl-9H-carbazole

B4795-1G B4795-5G
TCIAB4795-1GEA 39 EUR
TCIAB4795-1G TCIAB4795-5G
3-Bromo-6,9-diphenylcarbazole ≥98.0% (by GC)
3-Bromo-6,9-diphenylcarbazole
Formula: C₂₄H₁₆BrN
Peso molecolare: 398,30 g/mol
MDL Number: MFCD20257783
CAS Number: 1160294-85-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 143.0 to 147.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR