Stampa…

2-(3-Bromophenyl)-1-phenylbenzimidazole ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 2-(3-Bromophenyl)-1-phenyl-1H-benzoimidazole

B4794-1G B4794-5G
TCIAB4794-1GEA 58 EUR
TCIAB4794-1G TCIAB4794-5G
2-(3-Bromophenyl)-1-phenylbenzimidazole ≥98.0% (by GC, titration analysis)
2-(3-Bromophenyl)-1-phenylbenzimidazole
Formula: C₁₉H₁₃BrN₂
Peso molecolare: 349,23 g/mol
MDL Number: MFCD12911655
CAS Number: 760212-40-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light gray to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 123.0 to 127.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR