ethyl-2-((tert-butoxycarbonyl)amino)acetate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
N-(tert-Butoxycarbonyl)glycine ethyl ester, ethyl 2-((tert-Butoxycarbonyl)amino)acetate, Boc-Gly-OEt, N-Boc-glycine ethyl ester
B4782-5G
B4782-25G
TCIAB4782-5GEA
34
EUR
TCIAB4782-5G
TCIAB4782-25G
ethyl-2-((tert-butoxycarbonyl)amino)acetate ≥98.0% (by GC)
ethyl-2-((tert-butoxycarbonyl)amino)acetate
Formula:
C₉H₁₇NO₄ Peso molecolare: 203,2 g/mol Punto di ebollizione: 97°C |
MDL Number:
MFCD05663720 CAS Number: 14719-37-0 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...