4-Bromo-9,9-diphenylfluorene ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
4-Bromo-9,9-diphenyl-9H-fluorene
B4776-1G
B4776-5G
TCIAB4776-1GEA
49
EUR
TCIAB4776-1G
TCIAB4776-5G
4-Bromo-9,9-diphenylfluorene ≥98.0% (by GC)
4-Bromo-9,9-diphenylfluorene
Formula:
C₂₅H₁₇Br Peso molecolare: 397,32 g/mol |
MDL Number:
MFCD27923067 CAS Number: 713125-22-5 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 213.0 to 217.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...