Benzo[b]thiophene-3-boronic acid (contains varying amounts of Anhydride)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Benzothiophene-3-boronic acid, Benzo[b]thiophen-3-ylboronic acid
B4774-1G
B4774-5G
TCIAB4774-1GEA
33
EUR
TCIAB4774-1G
TCIAB4774-5G
Benzo[b]thiophene-3-boronic acid (contains varying amounts of Anhydride)
Benzo[b]thiophene-3-boronic acid
Formula:
C₈H₇BO₂S Peso molecolare: 178,02 g/mol Punto di fusione: 230°C |
MDL Number:
MFCD01114669 CAS Number: 113893-08-6 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...