4-Bromophenylboronic acid pinacol ester ≥98.0% (by GC)
Fornitore: TCI
Synonyms:
4-Bromophenylboronic acid, pinacol ester, 2-(4-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Bromophenylboronic acid Pinacol ester, 1-Bromo-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
Formula:
C₁₂H₁₆BBrO₂ |
MDL Number:
MFCD11110553 CAS Number: 68716-49-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Melting point | 69.0 to 73.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...