tert-Butyl-4-((tosyloxy)methyl)piperidine-1-carboxylate ≥98.0% (by total nitrogen basis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-Boc-4-(tosyloxymethyl)piperidine, 1-(tert-Butoxycarbonyl)-4-(tosyloxymethyl)piperidine, tert-Butyl 4-((tosyloxy)methyl)piperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-4-[(p-toluenesulphonyloxy)methyl]piperidine
B4759-5G
B4759-25G
TCIAB4759-5GEA
55
EUR
TCIAB4759-5G
TCIAB4759-25G
tert-Butyl-4-((tosyloxy)methyl)piperidine-1-carboxylate ≥98.0% (by total nitrogen basis)
tert-Butyl-4-((tosyloxy)methyl)piperidine-1-carboxylate
Formula:
C₁₈H₂₇NO₅S Peso molecolare: 369,48 g/mol |
MDL Number:
MFCD05864740 CAS Number: 166815-96-9 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 72.0 to 76.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...