3-Bromo-4-hydroxy-5-methoxybenzonitrile ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Formula:
C₈H₆BrNO₂ |
MDL Number:
MFCD01123027 CAS Number: 52805-45-5 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 144.0 to 148.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...