Stampa…

5-Bromo-2-chlorobenzaldehyde ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B4756-1G B4756-5G
TCIAB4756-1GEA 25 EUR
TCIAB4756-1G TCIAB4756-5G
5-Bromo-2-chlorobenzaldehyde ≥98.0% (by GC)
5-Bromo-2-chlorobenzaldehyde
Formula: C₇H₄BrClO
Peso molecolare: 219,46 g/mol
MDL Number: MFCD08445659
CAS Number: 189628-37-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR