Stampa…

4-(n-Butoxy)phenylacetic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 4-Butoxyphenylacetic acid

B4742-1G B4742-5G
TCIAB4742-1GEA 16 EUR
TCIAB4742-1G TCIAB4742-5G
4-(n-Butoxy)phenylacetic acid ≥98.0% (by GC, titration analysis)
4-(n-Butoxy)phenylacetic acid
Formula: C₁₂H₁₆O₃
Peso molecolare: 208.26 g/mol
MDL Number: MFCD00016832
CAS Number: 4547-57-3
EINECS: 224-904-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 85.0 to 89.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR