3-Bromo-5-hydroxybenzoic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
B4740-1G
B4740-5G
TCIAB4740-1GEA
37
EUR
TCIAB4740-1G
TCIAB4740-5G
3-Bromo-5-hydroxybenzoic acid ≥98.0% (by GC, titration analysis)
3-Bromo-5-hydroxybenzoic acid
Formula:
C₇H₅BrO₃ |
MDL Number:
MFCD06797980 CAS Number: 140472-69-1 UN: 3077 ADR: 9,III |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 237.0 to 241.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...