4-Benzyloxyphenylacetonitrile ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
2-(4-(Benzyloxy)phenyl)acetonitrile
Formula:
C₁₅H₁₃NO Punto di ebollizione: 173°C |
MDL Number:
MFCD00016400 CAS Number: 838-96-0 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...