Stampa…

4-(tert-Butoxycarbonylamino)cyclohexanol ≥98.0% (by GC, total nitrogen)

Fornitore: TCI

B4714-1G B4714-5G
TCIAB4714-1GEA 18 EUR
TCIAB4714-1G TCIAB4714-5G
4-(tert-Butoxycarbonylamino)cyclohexanol ≥98.0% (by GC, total nitrogen)
4-(tert-Butoxycarbonylamino)cyclohexanol
Formula: C₁₁H₂₁NO₃
MDL Number: MFCD06658349
CAS Number: 224309-64-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 170.0 to 174.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR