Benfotiamine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
Formula:
C₁₉H₂₃N₄O₆PS Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00057343 CAS Number: 22457-89-2 EINECS: 245-013-4 Merck Index: 14,01041 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...