1-Benzylpiperidine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
B4698-5ML
B4698-25ML
TCIAB4698-5MLEA
42
EUR
TCIAB4698-5ML
TCIAB4698-25ML
1-Benzylpiperidine ≥98.0% (by GC, titration analysis)
1-Benzylpiperidine
Formula:
C₁₂H₁₇N Peso molecolare: 175,28 g/mol Punto di ebollizione: 66°C |
MDL Number:
MFCD00224901 CAS Number: 2905-56-8 EINECS: 220-809-4 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...