Stampa…

6-Bromo-2-oxo-2H-chromene-3-carboxylic acid ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Bromocoumarin-3-carboxylic acid, UBP608

B4696-5G B4696-25G
TCIAB4696-5GEA 83 EUR
TCIAB4696-5G TCIAB4696-25G
6-Bromo-2-oxo-2H-chromene-3-carboxylic acid ≥98.0% (by HPLC)
6-Bromo-2-oxo-2H-chromene-3-carboxylic acid
Formula: C₁₀H₅BrO₄
Peso molecolare: 269,06 g/mol
Punto di fusione: 193°C
MDL Number: MFCD00047640
CAS Number: 2199-87-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR