Stampa…

1-Benzyloxy-4-iodobenzene ≥98.0% (by GC)

Fornitore: TCI

B4692-5G B4692-25G
TCIAB4692-5GEA 62 EUR
TCIAB4692-5G TCIAB4692-25G
1-Benzyloxy-4-iodobenzene ≥98.0% (by GC)
1-Benzyloxy-4-iodobenzene
Formula: C₁₃H₁₁IO
MDL Number: MFCD00079704
CAS Number: 19578-68-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 60.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR