3-(Bromoacetyl)-2H-chromen-2-one ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
3-(Bromoacetyl)coumarin, 3-(2-Bromoacetyl)-2H-chromen-2-one
B4679-1G
B4679-5G
TCIAB4679-1GEA
16
EUR
TCIAB4679-1G
TCIAB4679-5G
3-(Bromoacetyl)-2H-chromen-2-one ≥95.0% (by GC)
3-(Bromoacetyl)-2H-chromen-2-one
Formula:
C₁₁H₇BrO₃ Peso molecolare: 267,09 g/mol |
MDL Number:
MFCD00488331 CAS Number: 29310-88-1 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 163.0 to 167.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...