Stampa…

3-Bromo-2-hydroxy-5-nitropyridine ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Bromo-5-nitro-2(1H)-pyridinone, 3-Bromo-5-nitro-2-pyridinol, 3-Bromo-5-nitro-2-pyridone

B4672-5G B4672-25G
TCIAB4672-5GEA 47 EUR
TCIAB4672-5G TCIAB4672-25G
3-Bromo-2-hydroxy-5-nitropyridine ≥97.0% (by GC, titration analysis)
3-Bromo-2-hydroxy-5-nitropyridine
Formula: C5H3BrN2O3
Peso molecolare: 218,99 g/mol
MDL Number: MFCD03840431
CAS Number: 15862-33-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 214.0 to 218.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR