2-Bromophenanthrene ≥97.0% (by GC)
Fornitore: TCI
Formula:
C₁₄H₉Br Punto di ebollizione: 185°C |
MDL Number:
MFCD13191756 CAS Number: 62162-97-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 95.0 to 99.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...