4-Bromo-4',4''-dimethoxytriphenylamine ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI
|
Attenzione
|
Formula:
C₂₀H₁₈BrNO₂ |
MDL Number:
MFCD22124385 CAS Number: 194416-45-0 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...