Stampa…

2-Bromo-4,6-diphenylpyrimidine ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B4662-200MG B4662-1G
TCIAB4662-200MGEA 77 EUR
TCIAB4662-200MG TCIAB4662-1G
2-Bromo-4,6-diphenylpyrimidine ≥97.0% (by GC)
2-Bromo-4,6-diphenylpyrimidine
Formula: C₁₆H₁₁BrN₂
Peso molecolare: 311,18 g/mol
MDL Number: MFCD00234884
CAS Number: 56181-49-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 127.0 to 131.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR